MAT089756896 Matrix Scientific

1-(2-Fluoro-benzyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD07396656
  • Cas Number : 878733-50-7
  • Molecular Weight : 231.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019712.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019712.html
Availability details for
Quantity
  • Availability
  • Add to Cart