MAT089756903 Matrix Scientific

2-(2-Morpholin-4-yl-ethoxy)-phenylamine

  • MDL number MFCD04970982
  • Cas Number : 64039-56-1
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019719.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019719.html
Availability details for
Quantity
  • Availability
  • Add to Cart