MAT089756908 Matrix Scientific

Indan-5-carboxylic acid

  • MDL number MFCD04038659
  • Cas Number : 65898-38-6
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184
Country of Origin USA
Boiling point 126-128�6 Torr
Product Link http://www.matrixscientific.com/019724.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019724.html
Availability details for
Quantity
  • Availability
  • Add to Cart