MAT089756913 Matrix Scientific

2-(4-Fluoro-phenyl)-imidazo[1,2-a]pyrimidine-3-carbaldehyde

  • MDL number MFCD06653352
  • Cas Number : 878436-47-6
  • Molecular Weight : 241.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019729.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019729.html
Availability details for
Quantity
  • Availability
  • Add to Cart