MAT089756951 Matrix Scientific

3-(4-Fluoro-phenyl)-5-methyl-4,5-dihydro-isoxazole-5-carboxylic acid

  • MDL number MFCD06753283
  • Cas Number : 878426-96-1
  • Molecular Weight : 223.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019767.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019767.html
Availability details for
Quantity
  • Availability
  • Add to Cart