MAT089756985 Matrix Scientific

Piperidino-[2]piperidyl-methane

  • MDL number MFCD00085502
  • Cas Number : 81310-55-6
  • Molecular Weight : 182.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 104-106�Torr
Product Link http://www.matrixscientific.com/019801.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019801.html
Availability details for
Quantity
  • Availability
  • Add to Cart