MAT089756990 Matrix Scientific

1-Piperidino-2-[2]piperidyl-ethane

  • MDL number MFCD00083062
  • Cas Number : 14759-07-0
  • Molecular Weight : 196.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 107-108�2 Torr
Product Link http://www.matrixscientific.com/019806.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019806.html
Availability details for
Quantity
  • Availability
  • Add to Cart