MAT089756999 Matrix Scientific

1-(2-Ethoxy-phenyl)-5-methyl-1H-[1,2,3]triazole-4-carboxylic acid

  • MDL number MFCD07397773
  • Cas Number : 883538-73-6
  • Molecular Weight : 247.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019815.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019815.html
Availability details for
Quantity
  • Availability
  • Add to Cart