MAT089757003 Matrix Scientific

(4-Benzyl-piperidin-1-yl)-acetic acid

  • MDL number MFCD07186543
  • Cas Number : 438634-64-1
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-252
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/019819.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019819.html
Availability details for
Quantity
  • Availability
  • Add to Cart