MAT089757004 Matrix Scientific

Diisopropylamino-acetic acid

  • MDL number MFCD07189150
  • Cas Number : 44976-83-2
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-210
Country of Origin USA
Product Link http://www.matrixscientific.com/019820.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019820.html
Availability details for
Quantity
  • Availability
  • Add to Cart