MAT089757023 Matrix Scientific

3-(2-Methyl-indol-1-yl)-propylamine

  • MDL number MFCD06801051
  • Cas Number : 883542-35-6
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019839.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019839.html
Availability details for
Quantity
  • Availability
  • Add to Cart