MAT089757033 Matrix Scientific

C-(2,3,5-Trimethyl-1H-indol-7-yl)-methylamine

  • MDL number MFCD07186553
  • Cas Number : 883548-14-9
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019849.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019849.html
Availability details for
Quantity
  • Availability
  • Add to Cart