MAT089757042 Matrix Scientific

2-Ethyl-1-methyl-1H-benzoimidazol-5-ylamine

  • MDL number MFCD07186559
  • Cas Number : 323584-32-3
  • Molecular Weight : 175.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019858.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019858.html
Availability details for
Quantity
  • Availability
  • Add to Cart