MAT089757060 Matrix Scientific

1-(3,4-Dichloro-benzyl)-1H-pyrazol-4-ylamine

  • MDL number MFCD02056382
  • Cas Number : 895929-62-1
  • Molecular Weight : 242.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/019876.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019876.html
Availability details for
Quantity
  • Availability
  • Add to Cart