MAT089757075 Matrix Scientific

2-p-Tolyloxy-butyric acid

  • MDL number MFCD03422244
  • Cas Number : 143094-64-8
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Country of Origin USA
Product Link http://www.matrixscientific.com/019891.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019891.html
Availability details for
Quantity
  • Availability
  • Add to Cart