MAT089757080 Matrix Scientific

1-(2-Fluoro-benzyl)-5-methyl-1H-pyrazol-3-ylamine

  • MDL number MFCD04969964
  • Cas Number : 925146-07-2
  • Molecular Weight : 205.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019896.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019896.html
Availability details for
Quantity
  • Availability
  • Add to Cart