MAT089757092 Matrix Scientific

C-[1-(4-Methoxy-benzyl)-piperidin-4-yl]-methylamine

  • MDL number MFCD06809623
  • Cas Number : 117702-38-2
  • Molecular Weight : 234.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/019908.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/019908.html
Availability details for
Quantity
  • Availability
  • Add to Cart