MAT089757103 Matrix Scientific

4-(4-Methoxy-phenoxymethyl)-piperidine

  • MDL number MFCD07402197
  • Cas Number : 63608-38-8
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019919.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019919.html
Availability details for
Quantity
  • Availability
  • Add to Cart