MAT089757109 Matrix Scientific

1-Ethyl-piperidin-3-ylamine

  • MDL number MFCD00044587
  • Cas Number : 6789-94-2
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 155
Product Link http://www.matrixscientific.com/019925.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019925.html
Availability details for
Quantity
  • Availability
  • Add to Cart