MAT089757114 Matrix Scientific

1-(2-Chloro-ethoxy)-4-methyl-benzene

  • MDL number MFCD00634281
  • Cas Number : 20764-35-6
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-237
Country of Origin USA
Boiling point 114-116� Torr
Product Link http://www.matrixscientific.com/019930.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019930.html
Availability details for
Quantity
  • Availability
  • Add to Cart