MAT089757183 Matrix Scientific

2,4,5,6,7,8-Hexahydro-cycloheptapyrazole-3-carboxylic acid

  • MDL number MFCD06011089
  • Cas Number : 842972-72-9
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020004.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020004.html
Availability details for
Quantity
  • Availability
  • Add to Cart