MAT089757184 Matrix Scientific

6-Amino-3-methyl-3H-benzooxazol-2-one

  • MDL number MFCD07801206
  • Cas Number : 99584-10-8
  • Molecular Weight : 164.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020005.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020005.html
Availability details for
Quantity
  • Availability
  • Add to Cart