MAT089757192 Matrix Scientific

1-(2-Hydroxy-ethyl)-5-methyl-1H-benzoimidazole-2-carboxylic acid

  • MDL number MFCD07790315
  • Cas Number : 883542-66-3
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020013.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020013.html
Availability details for
Quantity
  • Availability
  • Add to Cart