MAT089757194 Matrix Scientific

4-(2-Fluoro-benzyloxy)-phenylamine

  • MDL number MFCD08146647
  • Cas Number : 57181-83-6
  • Molecular Weight : 217.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/020015.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020015.html
Availability details for
Quantity
  • Availability
  • Add to Cart