MAT089757195 Matrix Scientific

Hexafluoroisopropyl trifluoroacetate

  • MDL number MFCD08146653
  • Cas Number : 42031-15-2
  • Molecular Weight : 264.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020016.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020016.html
Availability details for
Quantity
  • Availability
  • Add to Cart