MAT089757196 Matrix Scientific

2-Chloro-N-[4-(3-methyl-piperidin-1-yl)-phenyl]-acetamide

  • MDL number MFCD06589845
  • Cas Number : 842974-39-4
  • Molecular Weight : 266.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019357.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019357.html
Availability details for
Quantity
  • Availability
  • Add to Cart