MAT089757217 Matrix Scientific

6-Amino-2,4-dimethyl-4H-benzo[1,4]oxazin-3-one

  • MDL number MFCD07788350
  • Cas Number : 70488-69-6
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020040.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020040.html
Availability details for
Quantity
  • Availability
  • Add to Cart