MAT089757242 Matrix Scientific

4-(5-Ethyl-2-methoxy-phenyl)-thiazol-2-ylamine

  • MDL number MFCD02663925
  • Cas Number : 383132-58-9
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020074.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020074.html
Availability details for
Quantity
  • Availability
  • Add to Cart