MAT089757254 Matrix Scientific

3-Dimethylaminomethyl-benzoic acid

  • MDL number MFCD07801107
  • Cas Number : 155412-73-0
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020089.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020089.html
Availability details for
Quantity
  • Availability
  • Add to Cart