MAT089757270 Matrix Scientific

4-Morpholin-4-yl-piperidine-4-carboxylic acidamide

  • MDL number MFCD07801137
  • Cas Number : 53654-12-9
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-186
Country of Origin USA
Product Link http://www.matrixscientific.com/020106.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020106.html
Availability details for
Quantity
  • Availability
  • Add to Cart