MAT089757322 Matrix Scientific

6-Amino-3H-benzothiazol-2-one

  • MDL number MFCD07792930
  • Cas Number : 56354-98-4
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221-222248
Country of Origin USA
Product Link http://www.matrixscientific.com/020159.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020159.html
Availability details for
Quantity
  • Availability
  • Add to Cart