MAT089757325 Matrix Scientific

4-(4-Allyl-5-hydroxy-3-methyl-pyrazol-1-yl)-benzoic acid

  • MDL number MFCD07793324
  • Cas Number : 1015844-62-8
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020162.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020162.html
Availability details for
Quantity
  • Availability
  • Add to Cart