MAT089757327 Matrix Scientific

4-Isobutoxy-phenylamine

  • MDL number MFCD01009861
  • Cas Number : 5198-04-9
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 145-146� Torr
Product Link http://www.matrixscientific.com/020164.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020164.html
Availability details for
Quantity
  • Availability
  • Add to Cart