MAT089757488 Matrix Scientific

Tricyclo[3.2.1.0*2,4*]octane-3-carboxylic acid

  • MDL number MFCD08056150
  • Cas Number : 37399-09-0
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020343.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020343.html
Availability details for
Quantity
  • Availability
  • Add to Cart