MAT089757508 Matrix Scientific

[1-(2-Chloro-phenyl)-5-hydroxy-3-methyl-1H-pyrazol-4-yl]-acetic acid

  • MDL number MFCD08056151
  • Cas Number : 1015844-43-5
  • Molecular Weight : 266.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020363.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020363.html
Availability details for
Quantity
  • Availability
  • Add to Cart