MAT089757532 Matrix Scientific

Isopropyl-(2-methoxy-ethyl)-amine

  • MDL number MFCD00144828
  • Cas Number : 104678-18-4
  • Molecular Weight : 117.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020388.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020388.html
Availability details for
Quantity
  • Availability
  • Add to Cart