MAT089757565 Matrix Scientific

2-Chloro-N-(4-methyl-6-oxo-1,6-dihydro-pyrimidin-2-yl)-acetamide

  • MDL number MFCD00457818
  • Cas Number : 341010-69-3
  • Molecular Weight : 201.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020418.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020418.html
Availability details for
Quantity
  • Availability
  • Add to Cart