MAT089757570 Matrix Scientific

N-(5-Amino-[1,2,4]thiadiazol-3-yl)-acetamide

  • MDL number MFCD02660455
  • Cas Number : 327059-94-9
  • Molecular Weight : 158.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020425.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020425.html
Availability details for
Quantity
  • Availability
  • Add to Cart