MAT089757575 Matrix Scientific

(4-Methyl-furazan-3-yl)-acetic acid

  • MDL number MFCD06010348
  • Cas Number : 15323-69-0
  • Molecular Weight : 142.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Country of Origin USA
Product Link http://www.matrixscientific.com/020430.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020430.html
Availability details for
Quantity
  • Availability
  • Add to Cart