MAT089757577 Matrix Scientific

[2-(2-Chloro-acetylamino)-thiazol-5-yl]-acetic acid methyl ester

  • MDL number MFCD04343724
  • Cas Number : 869951-10-0
  • Molecular Weight : 248.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020433.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020433.html
Availability details for
Quantity
  • Availability
  • Add to Cart