MAT089757588 Matrix Scientific

4-(2-Pyrimidinyloxy)benzenecarboxylic acid

  • MDL number MFCD03617910
  • Cas Number : 855423-33-5
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020444.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020444.html
Availability details for
Quantity
  • Availability
  • Add to Cart