MAT089757608 Matrix Scientific

3-(6,7-Dihydro-1H-5,8-dioxa-1,3-diaza-cyclopenta[b]naphthalen-2-yl)-propionic acid

  • MDL number MFCD07801149
  • Cas Number : 876717-89-4
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020465.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020465.html
Availability details for
Quantity
  • Availability
  • Add to Cart