MAT089757634 Matrix Scientific

2-Ethyl-2H-pyrazole-3-carboxylic acid(3-amino-phenyl)-amide

  • MDL number MFCD05667125
  • Cas Number : 957261-69-7
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020495.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020495.html
Availability details for
Quantity
  • Availability
  • Add to Cart