MAT089757649 Matrix Scientific

(2-Methyl-6,7-dihydro-5,8-dioxa-1,3-diaza-cyclopenta[b]naphthalen-1-yl)-acetic acid

  • MDL number MFCD08056109
  • Cas Number : 879045-15-5
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020510.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020510.html
Availability details for
Quantity
  • Availability
  • Add to Cart