MAT089757662 Matrix Scientific

3-(3-Pyrrolidin-1-ylmethyl-indol-1-yl)-propionic acid

  • MDL number MFCD08056121
  • Cas Number : 883546-91-6
  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020523.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020523.html
Availability details for
Quantity
  • Availability
  • Add to Cart