MAT089757669 Matrix Scientific

5-Chloro-1,2,3,4-tetrahydro-quinolin-8-ol

  • MDL number MFCD03820335
  • Cas Number : 54810-34-3
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-117
Country of Origin USA
Product Link http://www.matrixscientific.com/020530.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020530.html
Availability details for
Quantity
  • Availability
  • Add to Cart