MAT089757709 Matrix Scientific

4-Amino-3-chloropyridine

  • MDL number MFCD03984473
  • Cas Number : 19798-77-7
  • Molecular Weight : 128.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-46
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/020571.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020571.html
Availability details for
Quantity
  • Availability
  • Add to Cart