MAT089757731 Matrix Scientific

(2-Methylpyridin-4-yl)boronic acid

  • MDL number MFCD03095364
  • Cas Number : 579476-63-4
  • Molecular Weight : 136.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Country of Origin USA
Product Link http://www.matrixscientific.com/020595.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/020595.html
Availability details for
Quantity
  • Availability
  • Add to Cart