MAT089757770 Matrix Scientific

Phenethyl-hydrazine

  • MDL number MFCD00047825
  • Cas Number : 51-71-8
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Boiling point 74
Product Link http://www.matrixscientific.com/020638.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/020638.html
Availability details for
Quantity
  • Availability
  • Add to Cart