MAT089757788 Matrix Scientific

C-[3-(4-Chloro-phenyl)-1H-pyrazol-4-yl]-methylamine

  • MDL number MFCD08146630
  • Cas Number : 1020052-19-0
  • Molecular Weight : 207.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020656.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020656.html
Availability details for
Quantity
  • Availability
  • Add to Cart